Please login to your online account to display your discounted pricing

3,5-Di-tert-butyl-o-benzoquinone, 98%, ACROS Organics™

$186.00 - $186.00

Chemical Identifiers

Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
CAS 3383-21-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H20O2
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
MDL Number MFCD00001647
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC169300010

  SDS
thermo fisher scientific
AC169300010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC169300050

  SDS
thermo fisher scientific
169300050
5g Each for $186.00
Description & Specifications

Specifications

Physical Form Needle-Like Crystals
Assay ≥97.5%
Formula Weight 220.31
Color Red to brownish
Quantity 1g
Melting Point 114° to 117°C
Fieser 03,78
CAS Max % 100.0
Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
Assay Percent Range 98%
Molecular Formula C14H20O2
Infrared Spectrum Authentic
Linear Formula [(CH3)3C]2C6H2(=O)2
MDL Number MFCD00001647
CAS 3383-21-9
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.