Please login to your online account to display your discounted pricing

3,5-Di-tert-butyl-o-benzoquinone, 98%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  169300050

 View more versions of this product

Catalog No. AC169300050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Needle-Like Crystals
Assay ≥97.5%
Formula Weight 220.31
Color Red to brownish
Quantity 5g
Fieser 03,78
Melting Point 114° to 117°C
CAS Max % 100.0
Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C14H20O2
CAS 3383-21-9
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Linear Formula [(CH3)3C]2C6H2(=O)2
MDL Number MFCD00001647
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
CAS 3383-21-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C14H20O2
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
MDL Number MFCD00001647
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents