Please login to your online account to display your discounted pricing

3,5-Di-tert-butyl-o-benzoquinone, 97%, Maybridge


Quantity 25g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  BTB10297EB

 View more versions of this product

Catalog No. BTB10297EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 220.31
Quantity 25g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
Packaging Amber glass bottle
Molecular Formula C14H20O2
CAS 3383-21-9
CAS Min % 96.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
Linear Formula [(CH3)3C]2C6H2(=O)2
MDL Number MFCD00001647

Specifications

Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
CAS 3383-21-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H20O2
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
MDL Number MFCD00001647
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.