Please login to your online account to display your discounted pricing

3,5-Di-tert-butyl-o-benzoquinone, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
CAS 3383-21-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C14H20O2
Linear Formula [(CH3)3C]2C6H2(=O)2
Formula Weight 220.31
MDL Number MFCD00001647
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB10297EB

  SDS
fisher scientific company
BTB10297EB
Amber glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 220.31
Quantity 25g
Assay 97%
CAS Max % 100.0
Packaging Amber glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Chemical Name or Material 3, 5-Di-tert-butyl-o-benzoquinone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C14H20O2
CAS 3383-21-9
CAS Min % 96.5
Linear Formula [(CH3)3C]2C6H2(=O)2
MDL Number MFCD00001647
Assay Percent Range 97%
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.