Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Di-tert-butyl-4-hydroxyphenylacetonitrile, 98%

$65.73 - $272.45

Chemical Identifiers

Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylacetonitrile
CAS 1611-07-0
Molecular Formula C16H23NO
Formula Weight 245.37
MDL Number MFCD00156139
Synonym 3,5-Di-tert-butyl-4-hydroxybenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1349303

  SDS
alfa aesar™
A1349303
1g Each for $65.73

AAA1349306

  SDS
alfa aesar™
A1349306
5g Each for $272.45
Description & Specifications

Specifications

Formula Weight 245.37
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylacetonitrile
Assay 98%
Molecular Formula C16H23NO
MDL Number MFCD00156139
CAS 1611-07-0
Synonym 3,5-Di-tert-butyl-4-hydroxybenzyl cyanide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 109° to 112°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.