Learn More
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1051409
Description
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
Specifications
3,5-Di-tert-butyl-4-hydroxycinnamic acid | |
22014-01-3 | |
C17H24O3 | |
10 g | |
3-3,5-di-tert-butyl-4-hydroxyphenyl acrylic acid, unii-k7zj1m9j5t, k7zj1m9j5t, 3,5-di-tert-butyl-4-hydroxycinnamic acid, 3,5-di-t-butyl-4-hydroxycinnamic acid, 3,5-di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 2e-3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-bis tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-ditert-butyl-4-hydroxyphenyl acrylic acid | |
CTYWXRDQWMRIIM-BQYQJAHWSA-N | |
(E)-3-(3,5-ditert-butyl-4-hydroxyphenyl)prop-2-enoic acid | |
689095 | |
98% |
predominantly trans | |
∼195°C (decomposition) | |
MFCD00017291 | |
3144594 | |
Soluble in water. | |
CC(C)(C)C1=CC(\C=C\C(O)=O)=CC(=C1O)C(C)(C)C | |
276.38 | |
276.37 |
Chemical Identifiers
22014-01-3 | |
276.38 | |
CTYWXRDQWMRIIM-BQYQJAHWSA-N | |
689095 | |
CC(C)(C)C1=CC(\C=C\C(O)=O)=CC(=C1O)C(C)(C)C |
C17H24O3 | |
MFCD00017291 | |
3-3,5-di-tert-butyl-4-hydroxyphenyl acrylic acid, unii-k7zj1m9j5t, k7zj1m9j5t, 3,5-di-tert-butyl-4-hydroxycinnamic acid, 3,5-di-t-butyl-4-hydroxycinnamic acid, 3,5-di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 2e-3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-bis tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-ditert-butyl-4-hydroxyphenyl acrylic acid | |
(E)-3-(3,5-ditert-butyl-4-hydroxyphenyl)prop-2-enoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only