Please login to your online account to display your discounted pricing

3-(5-Bromo-pyridin-3-yl)-[1,2,4]oxadiazole-5-carboxylic acid ethyl ester, ≥97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  MO07248CB

 View more versions of this product

Catalog No. MO07248CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 298.1
Quantity 250mg
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
CAS 850375-34-7
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-(5-Bromo-pyridin-3-yl)-[1, 2, 4]oxadiazole-5-carboxylic acid ethyl ester
Molecular Formula C10H8BrN3O3
Health Hazard 1 GHS Signal Word: Warning
Packaging Amber Glass Bottle
MDL Number MFCD06797495

Specifications

Chemical Name or Material 3-(5-Bromo-pyridin-3-yl)-[1, 2, 4]oxadiazole-5-carboxylic acid ethyl ester
CAS 850375-34-7
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H8BrN3O3
Formula Weight 298.1
MDL Number MFCD06797495
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Causes serious eye irritation.

SDS & Safety Documents