Please login to your online account to display your discounted pricing

Alfa Aesar™ 3',5'-Bis(trifluoromethyl)propiophenone, 97%

$38.95 - $38.95

Chemical Identifiers

Chemical Name or Material 3',5'-Bis(trifluoromethyl)propiophenone
CAS 85068-34-4
Molecular Formula C11H8F6O
Formula Weight 270.18
MDL Number MFCD00009911
Beilstein 7484754
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2016803

  SDS
alfa aesar™
B2016803
1g Each for $38.95
Description & Specifications

Specifications

Formula Weight 270.18
MDL Number MFCD00009911
Quantity 1g
Chemical Name or Material 3',5'-Bis(trifluoromethyl)propiophenone
Assay 97%
Molecular Formula C11H8F6O
CAS 85068-34-4
Beilstein 7484754
Boiling Point 72°C (7mmHg)
Flash Point 76°C (168°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.