Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)phenylhydrazine, 97%

$62.68 - $937.20

Chemical Identifiers

Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylhydrazine
CAS 886-35-1
Molecular Formula C8H6F6N2
Formula Weight 244.14
MDL Number MFCD00051606
Beilstein 1843213
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1497103

  SDS
alfa aesar™
A1497103
1g Each for $62.68

AAA1497106

  SDS
alfa aesar™
A1497106
5g Each for $232.04

AAA1497114

  SDS
alfa aesar™
A1497114
25g Each for $937.20
Description & Specifications

Specifications

Formula Weight 244.14
Molecular Formula C8H6F6N2
Melting Point 80° to 83°C
Health Hazard 1 Danger
Chemical Name or Material 3,5-Bis(trifluoromethyl)phenylhydrazine
Assay 97%
MDL Number MFCD00051606
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Beilstein 1843213
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 886-35-1

Applications
3,5-Bis(trifluoromethyl)phenylhydrazine is used to measure formaldehyde concentration.

Solubility
Sparingly Soluble is water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.