missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)phenylhydrazine, 97%

Manufacturer:  Alfa Aesar™ A1497114

 View more versions of this product

Catalog No. AAA1497114



Description

Description

Applications
3,5-Bis(trifluoromethyl)phenylhydrazine is used to measure formaldehyde concentration.

Solubility
Sparingly Soluble is water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3,5-Bis(trifluoromethyl)phenylhydrazine
25g
C8H6F6N2
1843213
OULWACKZBGBRNT-UHFFFAOYSA-N
[3,5-bis(trifluoromethyl)phenyl]hydrazine
522901
97%
80°C to 83°C
886-35-1
MFCD00051606
3,5-bis trifluoromethyl phenylhydrazine, 3,5-bis-trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 3,5-di trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 1-3,5-bis trifluoromethyl phenyl hydrazine, hydrazine, 3,5-bis trifluoromethyl phenyl, pubchem1080, pubchem3289, timtec-bb sbb028449
C1=C(C=C(C=C1C(F)(F)F)NN)C(F)(F)F
244.14
244.14

Chemical Identifiers

886-35-1
244.14
OULWACKZBGBRNT-UHFFFAOYSA-N
522901
C1=C(C=C(C=C1C(F)(F)F)NN)C(F)(F)F
C8H6F6N2
MFCD00051606
3,5-bis trifluoromethyl phenylhydrazine, 3,5-bis-trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 3,5-di trifluoromethyl phenylhydrazine, 3,5-bis trifluoromethyl phenyl hydrazine, 1-3,5-bis trifluoromethyl phenyl hydrazine, hydrazine, 3,5-bis trifluoromethyl phenyl, pubchem1080, pubchem3289, timtec-bb sbb028449
[3,5-bis(trifluoromethyl)phenyl]hydrazine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents