missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane, 95%

Manufacturer:  Alfa Aesar™ L13709MD

 View more versions of this product

Catalog No. AAL13709MD



Specifications

Specifications

3,5-Bis(trifluoromethyl)phenyldimethylchlorosilane
C10H9ClF6Si
UN3265
JZWDIKPZWMXPMG-UHFFFAOYSA-N
[3,5-bis(trifluoromethyl)phenyl]-chloro-dimethylsilane
2783240
95%
250mg
732306-23-9
MFCD01862186
3,5-bis trifluoromethyl phenyldimethylchlorosilane, 3,5-bis trifluoromethyl phenyl chloro dimethylsilane, 3,5-bis trifluoromethyl phenyl chloro dimethyl silane, 3,5-bis trifluoromethyl phenyl-chloro-dimethylsilane, 3,5-bis trifluoromethyl phenyl dimethylsilyl chloride, benzene,1-chlorodimethylsilyl-3,5-bis trifluoromethyl
C[Si](C)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)Cl
306.707
306.71
95°C to 96°C (30mmHg)

Chemical Identifiers

732306-23-9
306.707
JZWDIKPZWMXPMG-UHFFFAOYSA-N
2783240
C[Si](C)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)Cl
C10H9ClF6Si
MFCD01862186
3,5-bis trifluoromethyl phenyldimethylchlorosilane, 3,5-bis trifluoromethyl phenyl chloro dimethylsilane, 3,5-bis trifluoromethyl phenyl chloro dimethyl silane, 3,5-bis trifluoromethyl phenyl-chloro-dimethylsilane, 3,5-bis trifluoromethyl phenyl dimethylsilyl chloride, benzene,1-chlorodimethylsilyl-3,5-bis trifluoromethyl
[3,5-bis(trifluoromethyl)phenyl]-chloro-dimethylsilane
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CHLOROSILANES, CORROSIVE, N.O.S.

Documents