missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Bis(trifluoromethyl)phenylacetic Acid, TCI America™

$59.58 - $189.45

Chemical Identifiers

CAS 85068-33-3
Molecular Formula C10H6F6O2
Molecular Weight (g/mol) 272.146
MDL Number MFCD00009908
InChI Key PAWSKKHEEYTXSA-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl phenylacetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 3,5-bis-trifluoromethylphenylacetic acid, mbt-paa, 2-3,5-di trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl acetic acid, 3,5-di trifluoromethyl phenylacetic acid, 3,5-bis trifluoromethyl phenylaceticacid
PubChem CID 123587
IUPAC Name 2-[3,5-bis(trifluoromethyl)phenyl]acetic acid
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B19761G View Documents TCI America
B19761G
1g Each for $59.58
 
B19765G View Documents TCI America
B19765G
5g Each for $189.45
 
Specifications

Chemical Identifiers

85068-33-3
272.146
PAWSKKHEEYTXSA-UHFFFAOYSA-N
123587
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CC(=O)O
C10H6F6O2
MFCD00009908
3,5-bis trifluoromethyl phenylacetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 3,5-bis-trifluoromethylphenylacetic acid, mbt-paa, 2-3,5-di trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl acetic acid, 3,5-di trifluoromethyl phenylacetic acid, 3,5-bis trifluoromethyl phenylaceticacid
2-[3,5-bis(trifluoromethyl)phenyl]acetic acid

Specifications

White-Yellow
1g
C10H6F6O2
3,5-bis trifluoromethyl phenylacetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 2-3,5-bis trifluoromethyl phenyl acetic acid, 3,5-bis-trifluoromethylphenylacetic acid, mbt-paa, 2-3,5-di trifluoromethyl phenyl acetic acid, benzeneacetic acid, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl acetic acid, 3,5-di trifluoromethyl phenylacetic acid, 3,5-bis trifluoromethyl phenylaceticacid
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CC(=O)O
272.146
272.15
≥97.0% (T)
125°C
85068-33-3
MFCD00009908
PAWSKKHEEYTXSA-UHFFFAOYSA-N
2-[3,5-bis(trifluoromethyl)phenyl]acetic acid
123587
Crystal-Powder at 20°C
3,5-Bis(trifluoromethyl)phenylacetic Acid
Safety and Handling

Safety and Handling

TSCA : No