Please login to your online account to display your discounted pricing

3,5-Bis(trifluoromethyl)phenylacetic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Bis(trifluoromethyl)phenylacetic acid
CAS 85068-33-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H6F6O2
Linear Formula (CF3)C6H3CHCOH
Formula Weight 272.14
MDL Number MFCD00009908
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303650050

  SDS
thermo fisher scientific
AC303650050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 272.14
Quantity 5g
Color White to beige
Melting Point 124° to 128°C
CAS Max % 100.0
Chemical Name or Material 3, 5-Bis(trifluoromethyl)phenylacetic acid
Packaging Glass bottle
Molecular Formula C10H6F6O2
MDL Number MFCD00009908
CAS 85068-33-3
CAS Min % 97.5
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (CF3)C6H3CHCOH
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.