missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Bis(trifluoromethyl)phenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1018314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,5-Bis (trifluoromethyl)phenol | |
20°C to 21°C | |
97°C to 98°C (50 mmHg) | |
C8H4F6O | |
MFCD00000386 | |
2123670 | |
ODSXJQYJADZFJX-UHFFFAOYSA-N | |
3,5-bis(trifluoromethyl)phenol | |
67680 | |
97% |
349-58-6 | |
1.511 | |
>110°C (230°F) | |
1.415 | |
25 g | |
3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr | |
C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F | |
230.109 | |
230.11 |
Chemical Identifiers
349-58-6 | |
230.109 | |
ODSXJQYJADZFJX-UHFFFAOYSA-N | |
67680 | |
C1=C(C=C(C=C1C(F)(F)F)O)C(F)(F)F |
C8H4F6O | |
MFCD00000386 | |
3,5-bis trifluoromethyl phenol, phenol, 3,5-bis trifluoromethyl, 3,5-di trifluoromethyl phenol, 3,5-bis-trifluoromethylphenol, 3,5-ditrifluoromethylphenol, 3,5-bis-trifluoromethyl phenol, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-3,5-xylenol, pubchem12483, mbt-oh, acmc-1clsr | |
3,5-bis(trifluoromethyl)phenol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-488-3
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only