Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)cinnamonitrile, 98%

$32.69 - $403.68

Chemical Identifiers

Chemical Name or Material 3,5-Bis (trifluoromethyl)cinnamonitrile
CAS 175136-63-7
Molecular Formula C11H5F6N
Formula Weight 265.16g/mol
MDL Number MFCD00051605
Synonym 3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1127903

 
alfa aesar™
A11279-03
1g Each for $32.69

AAA1127906

 
alfa aesar™
A11279-06
5g Each for $403.68
Description & Specifications

Specifications

Melting Point 80° to 83°C
Quantity 1g
Chemical Name or Material 3,5-Bis (trifluoromethyl)cinnamonitrile
CAS 175136-63-7
Assay 98%
Molecular Formula C11H5F6N
Formula Weight 265.16g/mol
MDL Number MFCD00051605
Synonym 3-[3,5-Bis(trifluoromethyl)phenyl]acrylonitrile
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for sev

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.