missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Bis(trifluoromethyl)bromobenzene, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 307700500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3, 5-Bis(trifluoromethyl)bromobenzene | |
1.7100g/mL | |
>110°C | |
98.5% min. (GC) | |
C8H3BrF6 | |
1.4260 to 1.4280 | |
1.71 | |
CSVCVIHEBDJTCJ-UHFFFAOYSA-N | |
1-bromo-3,5-bis(trifluoromethyl)benzene | |
67602 | |
99% |
328-70-1 | |
154.0°C | |
Authentic | |
Glass bottle | |
(CF3)2C6H3Br | |
50g | |
3,5-bis trifluoromethyl bromobenzene, 1-bromo-3,5-bis trifluoromethyl benzene, 1,3-bis trifluoromethyl-5-bromobenzene, mbt-br, 3,5-di trifluoromethyl bromobenzene, 3,5-bis trifluoromethyl-1-bromobenzene, 3,5-bis-trifluoromethylbromobenzene, benzene, 1-bromo-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenyl bromide, 1-bromo-3,5-bis-trifluoromethyl-benzene | |
C1=C(C=C(C=C1C(F)(F)F)Br)C(F)(F)F | |
293.01 | |
293.01 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 206-334-5
RUO â Research Use Only