Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl chloride
CAS 75462-59-8
Molecular Formula C9H5ClF6
Formula Weight 262.58
MDL Number MFCD00009906
UN Number UN3261
Beilstein 656502
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1655303

 
alfa aesar™
A1655303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1655306

 
alfa aesar™
A1655306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.58
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl chloride
Assay 97%
CAS 75462-59-8
Beilstein 656502
Melting Point 29° to 33°C
Boiling Point 46°C (3mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Molecular Formula C9H5ClF6
MDL Number MFCD00009906
UN Number UN3261
Quantity 1g
Flash Point 71°C (161°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3,5-Bis(trifluoromethyl)benzyl chloride has been used in the preparation of 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene. 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene, in 94% yield by an efficient nucleophilic substitution reaction between 3,5-bis-(trifluoromethyl)benzyl chloride, 4, and sodium azide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.