Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzyl bromide, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl bromide
CAS 32247-96-4
Molecular Formula C9H5BrF6
Formula Weight 307.04
MDL Number MFCD00009905
UN Number UN3265
Beilstein 656506
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1959003

  SDS
alfa aesar™
A1959003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1959006

  SDS
alfa aesar™
A1959006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1959014

  SDS
alfa aesar™
A1959014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.04
UN Number UN3265
CAS 32247-96-4
MDL Number MFCD00009905
Assay 98%
Health Hazard 1 Danger
Boiling Point 136° to 140°C (14mmHg)
Quantity 1g
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl bromide
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Beilstein 656506
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point >100°C (212°F)
Molecular Formula C9H5BrF6

Applications
3,5-Bis(trifluoromethyl)benzyl bromide is used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry in enantioselective synthesis of non-peptidic neurokinin NK1 receptor antagonist.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.