missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Bis(trifluoromethyl)benzyl Alcohol, TCI America™

$126.81 - $381.95

Chemical Identifiers

CAS 32707-89-4
Molecular Formula C9H6F6O
Molecular Weight (g/mol) 244.136
MDL Number MFCD00009907
InChI Key BJTWPJOGDWRYDD-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl alcohol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl-phenyl-methanol, 3,5-ditrifluoromethylbenzyl alcohol, benzenemethanol, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylalcohol, 3,5-di trifluoromethyl benzyl alcohol, 3,5-di trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl benzyl alcohol
PubChem CID 122933
IUPAC Name [3,5-bis(trifluoromethyl)phenyl]methanol
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17525G View Documents TCI America
B17525G
5g Each for $126.81
 
B175225G View Documents TCI America
B175225G
25g Each for $381.95
 
Specifications

Chemical Identifiers

32707-89-4
244.136
BJTWPJOGDWRYDD-UHFFFAOYSA-N
122933
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CO
C9H6F6O
MFCD00009907
3,5-bis trifluoromethyl benzyl alcohol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl-phenyl-methanol, 3,5-ditrifluoromethylbenzyl alcohol, benzenemethanol, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylalcohol, 3,5-di trifluoromethyl benzyl alcohol, 3,5-di trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl benzyl alcohol
[3,5-bis(trifluoromethyl)phenyl]methanol

Specifications

White
5g
C9H6F6O
3,5-bis trifluoromethyl benzyl alcohol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl-phenyl-methanol, 3,5-ditrifluoromethylbenzyl alcohol, benzenemethanol, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzylalcohol, 3,5-di trifluoromethyl benzyl alcohol, 3,5-di trifluoromethyl phenyl methanol, 3,5-bis-trifluoromethyl benzyl alcohol
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CO
244.136
244.14
≥98.0% (GC)
55°C
32707-89-4
MFCD00009907
BJTWPJOGDWRYDD-UHFFFAOYSA-N
[3,5-bis(trifluoromethyl)phenyl]methanol
122933
Crystal-Powder at 20°C
3,5-Bis(trifluoromethyl)benzyl Alcohol
Safety and Handling

Safety and Handling

TSCA : No