Learn More
3,5-Bis(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
$91.44 - $472.18
Chemical Identifiers
CAS | 785-56-8 |
---|---|
Molecular Formula | C9H3ClF6O |
Molecular Weight (g/mol) | 276.562 |
MDL Number | MFCD00000387 |
InChI Key | WAKMMQSMEDJRRI-UHFFFAOYSA-N |
Synonym | 3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride |
PubChem CID | 101856 |
IUPAC Name | 3,5-bis(trifluoromethyl)benzoyl chloride |
SMILES | C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1140106
|
Thermo Scientific Chemicals
A1140106 |
5 g |
Each for $91.44
|
|
|||||
AAA1140114
|
Thermo Scientific Chemicals
A1140114 |
25 g |
Each for $472.18
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
785-56-8 | |
276.562 | |
WAKMMQSMEDJRRI-UHFFFAOYSA-N | |
101856 | |
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl |
C9H3ClF6O | |
MFCD00000387 | |
3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride | |
3,5-bis(trifluoromethyl)benzoyl chloride |
Specifications
785-56-8 | |
65°C to 67°C (12 mmHg) | |
C9H3ClF6O | |
MFCD00000387 | |
UN3265 | |
Moisture sensitive | |
WAKMMQSMEDJRRI-UHFFFAOYSA-N | |
3,5-bis(trifluoromethyl)benzoyl chloride | |
101856 | |
97% |
1.526 | |
72°C (161°F) | |
1.4332 | |
5 g | |
2593440 | |
3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride | |
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl | |
276.562 | |
276.57 | |
3,5-Bis (trifluoromethyl)benzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 212-322-0
TSCA : No
Recommended Storage : Ambient temperatures