Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzophenone, 95%

$53.26 - $829.68

Chemical Identifiers

Chemical Name or Material 3,5-Bis(trifluoromethyl)benzophenone
CAS 21221-93-2
Molecular Formula C15H8F6O
Formula Weight 318.22
MDL Number MFCD00015510
Beilstein 3066566
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2004603

 
alfa aesar™
B2004603
1g Each for $53.26

AAB2004606

 
alfa aesar™
B2004606
5g Each for $197.03

AAB2004614

 
alfa aesar™
B2004614
25g Each for $829.68
Description & Specifications

Specifications

Formula Weight 318.22
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzophenone
Assay 95%
Molecular Formula C15H8F6O
MDL Number MFCD00015510
CAS 21221-93-2
Beilstein 3066566
Boiling Point 111° to 112°C (0.2mmHg)
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.