missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Bis(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1546014

 View more versions of this product

Catalog No. AAA1546014



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3,5-Bis(trifluoromethyl)benzonitrile
16°C to 20°C
154°C to 156°C
C9H3F6N
MFCD00000379
UN3276
3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene
FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F
239.12
239.12
27126-93-8
1.421
72°C (161°F)
1.4175
25 g
3552650
CZKHHAOIHXHOSR-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzonitrile
96746
98%

Chemical Identifiers

27126-93-8
239.12
CZKHHAOIHXHOSR-UHFFFAOYSA-N
96746
FC(F)(F)C1=CC(=CC(=C1)C#N)C(F)(F)F
C9H3F6N
MFCD00000379
3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene
3,5-bis(trifluoromethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H312-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c

H227-H302+H312+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

EINECSNumber : 248-240-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only