missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzonitrile, 98%

Manufacturer:  Alfa Aesar™ A1546003

 View more versions of this product

Catalog No. AAA1546003



Specifications

Specifications

3,5-Bis(trifluoromethyl)benzonitrile
154°C to 156°C
1g
C9H3F6N
UN3276
3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C#N
239.12
239.12
72°C (161°F)
16°C to 20°C
27126-93-8
MFCD00000379
3552650
CZKHHAOIHXHOSR-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzonitrile
96746
98%

Chemical Identifiers

27126-93-8
239.12
CZKHHAOIHXHOSR-UHFFFAOYSA-N
96746
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C#N
C9H3F6N
MFCD00000379
3,5-bis trifluoromethyl benzonitrile, 3,5-di trifluoromethyl benzonitrile, bis 3,5-trifluoromethyl benzonitrile, mbt-cn, 3,5-bis-trifluoromethyl benzonitrile, benzonitrile, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl benzenecarbonitrile, fxffr ccn exfff, pubchem4691, 3,5-bis tifluoromethyl-1-cyanobenzene
3,5-bis(trifluoromethyl)benzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H312-H332-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Documents