Learn More
3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1030003
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3,5-Bis (trifluoromethyl)benzenesulfonyl chloride | |
34°C to 38°C | |
MFCD00014725 | |
UN3261 | |
Moisture sensitive | |
BTRCVKADYDVSLI-UHFFFAOYSA-N | |
3,5-bis(trifluoromethyl)benzenesulfonyl chloride | |
520945 | |
97% |
39234-86-1 | |
C8H3ClF6O2S | |
1 g | |
3621218 | |
3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724 | |
C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F | |
312.61 | |
312.61 |
Chemical Identifiers
39234-86-1 | |
312.61 | |
BTRCVKADYDVSLI-UHFFFAOYSA-N | |
520945 | |
C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F |
C8H3ClF6O2S | |
MFCD00014725 | |
3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724 | |
3,5-bis(trifluoromethyl)benzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 254-371-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only