missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Bis(trifluoromethyl)benzeneboronic acid, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 351360050

 View more versions of this product

Catalog No. AC351360050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Authentic
White to Yellow
5g
73852-19-4
100.0
C8H5BF6O2
3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boranediol, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis trifluoromethyl phenylboroic acid, 3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, btfpba
B(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(O)O
257.93
257.93
95%
Glass bottle
219°C to 226°C
95%
94.0
95%
MFCD00051850
BPTABBGLHGBJQR-UHFFFAOYSA-N
[3,5-bis(trifluoromethyl)phenyl]boronic acid
156265
Crystals or Powder

Chemical Identifiers

73852-19-4
257.93
BPTABBGLHGBJQR-UHFFFAOYSA-N
156265
B(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(O)O
C8H5BF6O2
MFCD00051850
3,5-bis trifluoromethyl benzeneboronic acid, 3,5-bis trifluoromethyl phenyl boranediol, 3,5-bis trifluoromethyl phenyl boronic acid, 3,5-bis trifluoromethyl phenylboroic acid, 3,5-bis trifluoromethyl phenylboronic acid, 3,5-bis trifluoromethylphenyl boronic acid, 3,5-bis-trifluoromethylphenylboronic acid, btfpba
[3,5-bis(trifluoromethyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents