Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzamide, 97%

$31.24 - $354.04

Chemical Identifiers

Chemical Name or Material 3,5-Bis(trifluoromethyl)benzamide
CAS 22227-26-5
Molecular Formula C9H5F6NO
Formula Weight 257.14
MDL Number MFCD00015511
Beilstein 4734142
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2391703

  SDS
alfa aesar™
B2391703
1g Each for $31.24

AAB2391706

  SDS
alfa aesar™
B2391706
5g Each for $103.59

AAB2391714

  SDS
alfa aesar™
B2391714
25g Each for $354.04
Description & Specifications

Specifications

Formula Weight 257.14
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzamide
Assay 97%
Molecular Formula C9H5F6NO
MDL Number MFCD00015511
CAS 22227-26-5
Beilstein 4734142
Melting Point 161° to 163°C
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.