missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,5-Bis(trifluoromethyl)benzaldehyde 95.0+%, TCI America™
$31.52 - $122.76
Chemical Identifiers
CAS | 401-95-6 |
---|---|
Molecular Formula | C9H4F6O |
Molecular Weight (g/mol) | 242.12 |
MDL Number | MFCD00010206 |
InChI Key | LDWLIXZSDPXYDR-UHFFFAOYSA-N |
Synonym | 3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j |
PubChem CID | 136247 |
IUPAC Name | 3,5-bis(trifluoromethyl)benzaldehyde |
SMILES | C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O |
Chemical Identifiers
401-95-6 | |
242.12 | |
LDWLIXZSDPXYDR-UHFFFAOYSA-N | |
136247 | |
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O |
C9H4F6O | |
MFCD00010206 | |
3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j | |
3,5-bis(trifluoromethyl)benzaldehyde |
Specifications
401-95-6 | |
171°C | |
MFCD00010206 | |
3,5-bis trifluoromethyl benzaldehyde, 3,5-di trifluoromethyl benzaldehyde, mbt-bad, benzaldehyde, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl-benzaldehyde, 3,5-bis-trifluoromethyl benzaldehyde, pubchem4162, acmc-1ajhv, ksc490m1j | |
C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C=O | |
242.12 | |
242.12 | |
Liquid |
Yellow | |
C9H4F6O | |
5 g | |
LDWLIXZSDPXYDR-UHFFFAOYSA-N | |
3,5-bis(trifluoromethyl)benzaldehyde | |
136247 | |
≥95.0% (GC) | |
3,5-Bis(trifluoromethyl)benzaldehyde |
Safety and Handling
TSCA : No