Please login to your online account to display your discounted pricing

3,5-Bis(trifluoromethyl)benzaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzaldehyde
CAS 401-95-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H4F6O
Linear Formula (CF3)C6H3CHO
Formula Weight 242.12
MDL Number MFCD00010206
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC307690250

  SDS
thermo fisher scientific
AC307690250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC307690010

  SDS
thermo fisher scientific
AC307690010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC307690050

  SDS
thermo fisher scientific
307690050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 242.12
Quantity 25g
Color Clear colorless to yellow
Density 1.47g/mL
CAS Min % 96.0
CAS Max % 100.0
MDL Number MFCD00010206
CAS 401-95-6
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4215 to 1.4235 (20°C, 589nm)
Molecular Formula C9H4F6O
Linear Formula (CF3)C6H3CHO
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzaldehyde
Assay Percent Range 97%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.