Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(trifluoromethyl)benzaldehyde, 97%

$36.01 - $503.26

Chemical Identifiers

Chemical Name or Material 3,5-Bis(trifluoromethyl)benzaldehyde
CAS 401-95-6
Molecular Formula C9H4F6O
Formula Weight 242.12
MDL Number MFCD00010206
Beilstein 1884058
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1825903

  SDS
alfa aesar™
A1825903
1g Each for $36.01

AAA1825906

  SDS
alfa aesar™
A1825906
5g Each for $123.24

AAA1825914

  SDS
alfa aesar™
A1825914
25g Each for $503.26
Description & Specifications

Specifications

Formula Weight 242.12
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzaldehyde
Assay 97%
Molecular Formula C9H4F6O
MDL Number MFCD00010206
CAS 401-95-6
Beilstein 1884058
Boiling Point 171°C
Quantity 1g
Flash Point 70°C (158°F)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3,5-Bis(trifluoromethyl)benzaldehyde was used in the synthesis of a series of meso-3,5-bis(trifluoromethyl)phenyl-substituted expanded porphyrins.

Solubility
Insoluble in water. Soluble in many organic solvents.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.