missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3',5'-Bis(trifluoromethyl)acetanilide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals A1767506

 View more versions of this product

Catalog No. AAA1767506



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3',5'-Bis(trifluoromethyl)acetanilide
160°C to 162°C
MFCD00089454
3',5'-bis trifluoromethyl acetanilide, n-3,5-bis trifluoromethyl phenyl acetamide, 3,5-bis trifluoromethyl acetanilide, 3,5-dicf3 acetanilide, gnf-pf-1581, 1-acetamido-3,5-bis trifluoromethyl benzene, n1-3,5-di trifluoromethyl phenyl acetamide, maybridge1_000017, cbmicro_045192
CC(=O)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
271.162
271.16
16143-84-3
C10H7F6NO
5 g
XMKZELXFBZUGEY-UHFFFAOYSA-N
N-[3,5-bis(trifluoromethyl)phenyl]acetamide
723600
98%

Chemical Identifiers

16143-84-3
271.162
XMKZELXFBZUGEY-UHFFFAOYSA-N
723600
CC(=O)NC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
C10H7F6NO
MFCD00089454
3',5'-bis trifluoromethyl acetanilide, n-3,5-bis trifluoromethyl phenyl acetamide, 3,5-bis trifluoromethyl acetanilide, 3,5-dicf3 acetanilide, gnf-pf-1581, 1-acetamido-3,5-bis trifluoromethyl benzene, n1-3,5-di trifluoromethyl phenyl acetamide, maybridge1_000017, cbmicro_045192
N-[3,5-bis(trifluoromethyl)phenyl]acetamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions