missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5-Bis(tert-butylthio)-1-chlorobenzene, TCI America™

$212.36 - $627.92

Chemical Identifiers

CAS 260968-02-3
Molecular Formula C14H21ClS2
Molecular Weight (g/mol) 288.892
InChI Key QVEUHOXOWGXTJG-UHFFFAOYSA-N
Synonym 1-Chloro-3,5-bis(tert-butylthio)benzene
PubChem CID 22617204
IUPAC Name 1,3-bis(tert-butylsulfanyl)-5-chlorobenzene
SMILES CC(C)(C)SC1=CC(=CC(=C1)Cl)SC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B26591G View Documents TCI America
B26591G
1g Each for $212.36
 
B26595G View Documents TCI America
B26595G
5g Each for $627.92
 
Specifications

Chemical Identifiers

260968-02-3
288.892
1-Chloro-3,5-bis(tert-butylthio)benzene
1,3-bis(tert-butylsulfanyl)-5-chlorobenzene
C14H21ClS2
QVEUHOXOWGXTJG-UHFFFAOYSA-N
22617204
CC(C)(C)SC1=CC(=CC(=C1)Cl)SC(C)(C)C

Specifications

White-Yellow
260968-02-3
1-Chloro-3,5-bis(tert-butylthio)benzene
CC(C)(C)SC1=CC(=CC(=C1)Cl)SC(C)(C)C
288.892
288.89
≥98.0% (GC)
1g
C14H21ClS2
QVEUHOXOWGXTJG-UHFFFAOYSA-N
1,3-bis(tert-butylsulfanyl)-5-chlorobenzene
22617204
Crystal-Powder at 20°C
3,5-Bis(tert-butylthio)-1-chlorobenzene
Safety and Handling

Safety and Handling

TSCA : No