Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5-Bis(bromomethyl)toluene, 98%

Chemical Identifiers

Chemical Name or Material 3,5-Bis(bromomethyl)toluene
CAS 19294-04-3
Molecular Formula C9H10Br2
Formula Weight 277.99
MDL Number MFCD00182480
UN Number UN1759
Synonym alpha,alpha'-Dibromomesitylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1800209

  SDS
alfa aesar™
A1800209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1800218

  SDS
alfa aesar™
A1800218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.99
Chemical Name or Material 3,5-Bis(bromomethyl)toluene
Assay 98%
Molecular Formula C9H10Br2
MDL Number MFCD00182480
CAS 19294-04-3
UN Number UN1759
Melting Point 64° to 67°C
Synonym alpha,alpha'-Dibromomesitylene
Quantity 10g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Applications
3,5-Bis(bromomethyl)toluene is used as an anastrazole intermediate for the preparation of medicines and other fine chemicals. It is used as letrozole intermediates which is an oral non-steroidal aromatase inhibitor for the treatment of hormonally-responsive breast cancer after surgery.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.