missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3,5,7-Trimethyladamantane-1-carboxylic acid, 97%, Maybridge

Manufacturer:  Fisher Scientific Company BTB12973DE

 View more versions of this product

Catalog No. BTB12973DE



Specifications

Specifications

3, 5, 7-Trimethyladamantane-1-carboxylic acid
5g
96.5
97%
MFCD00187581
DHIIUPVOLLOORM-UHFFFAOYSA-N
3,5,7-trimethyladamantane-1-carboxylic acid
2802268
97%
Amber Glass Bottle
15291-66-4
100.0
C14H22O2
1,3,5-trimethyladamantane-7-carboxylic acid
CC12CC3(CC(C1)(CC(C2)(C3)C(=O)O)C)C
222.328
222.33

Chemical Identifiers

15291-66-4
222.328
DHIIUPVOLLOORM-UHFFFAOYSA-N
2802268
CC12CC3(CC(C1)(CC(C2)(C3)C(=O)O)C)C
C14H22O2
MFCD00187581
1,3,5-trimethyladamantane-7-carboxylic acid
3,5,7-trimethyladamantane-1-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents