Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5,6-Trichlorosalicylic acid, 98%

$66.15 - $186.84

Chemical Identifiers

Chemical Name or Material 3,5,6-Trichlorosalicylic acid
CAS 40932-60-3
Molecular Formula C7H3Cl3O3
Formula Weight 241.46
MDL Number MFCD00075245
Beilstein 2940592
Synonym 3,5,6-Trichloro-2-hydroxybenzoic acid; 2-Hydroxy-3,5,6-trichlorobenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2045514

  SDS
alfa aesar™
B2045514
25g Each for $66.15

AAB2045522

  SDS
alfa aesar™
B2045522
100g Each for $186.84
Description & Specifications

Specifications

Formula Weight 241.46
Chemical Name or Material 3,5,6-Trichlorosalicylic acid
TSCA TSCA
Assay 98%
Molecular Formula C7H3Cl3O3
MDL Number MFCD00075245
CAS 40932-60-3
Beilstein 2940592
Synonym 3,5,6-Trichloro-2-hydroxybenzoic acid; 2-Hydroxy-3,5,6-trichlorobenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 209° to 211°C
Quantity 25g

Applications
3,5,6-Trichlorosalicylic acid is a chlorinated salicylic acid intermediate.

Solubility
It has limited solubility in water.

Notes
Desiccate at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.