Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5,5-Trimethyloxazolidine-2,4-dione, 98%

$59.57 - $564.92

Chemical Identifiers

Chemical Name or Material 3,5,5-Trimethyloxazolidine-2,4-dione
CAS 127-48-0
Molecular Formula C6H9NO3
Formula Weight 143.14
MDL Number MFCD00047084
Beilstein 121627
Merck Index 14,9706
Synonym Trimethadione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2436203

 
alfa aesar™
B2436203
1g Each for $59.57

AAB2436206

 
alfa aesar™
B2436206
5g Each for $153.47

AAB2436214

 
alfa aesar™
B2436214
25g Each for $564.92
Description & Specifications

Specifications

Formula Weight 143.14
Chemical Name or Material 3,5,5-Trimethyloxazolidine-2,4-dione
Assay 98%
Molecular Formula C6H9NO3
MDL Number MFCD00047084
CAS 127-48-0
Beilstein 121627
Merck Index 14,9706
Synonym Trimethadione
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number RQ2100000
Boiling Point 78° to 80°C (5mmHg)
Melting Point 45° to 48°C
Quantity 1g

Solubility
Soluble in water 50 g/L.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360
May damage fertility or the unborn child.