Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,5,5-Trimethylcyclohexane-1,2-dione, 99%

$227.81 - $227.81

Chemical Identifiers

Chemical Name or Material 3,5,5-Trimethylcyclohexane-1,2-dione
CAS 57696-89-6
Molecular Formula C9H14O2
Formula Weight 154.21
MDL Number MFCD02181023
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2262514

 
alfa aesar™
B2262514
25g Each for $227.81
Description & Specifications

Specifications

Formula Weight 154.21
Chemical Name or Material 3,5,5-Trimethylcyclohexane-1,2-dione
TSCA TSCA
Assay 99%
Molecular Formula C9H14O2
MDL Number MFCD02181023
CAS 57696-89-6
Flash Point >100°C (212°F)
Melting Point 89° to 92°C
Quantity 25g