Please login to your online account to display your discounted pricing

3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  MO01247DE

 View more versions of this product

Catalog No. MO01247DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 191.04
Quantity 5g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
MDL Number MFCD03425979
CAS 845885-86-1
Assay Percent Range 97%
Molecular Formula C10H14BNO2
Health Hazard 2 GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)pyridine
CAS 845885-86-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H14BNO2
Formula Weight 191.04
MDL Number MFCD03425979
Accessories

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents