missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-[4-(Trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H5401006

 View more versions of this product

Catalog No. AAH5401006


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-[4-(Trifluoromethyl)phenyl]propionic acid
99°C
MFCD00674032
4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
C1=CC(=CC=C1CCC(=O)O)C(F)(F)F
218.175
218.17
53473-36-2
C10H9F3O2
5 g
OEIUMLSCWINLBB-UHFFFAOYSA-N
3-[4-(trifluoromethyl)phenyl]propanoic acid
3588736
97%

Chemical Identifiers

53473-36-2
218.175
OEIUMLSCWINLBB-UHFFFAOYSA-N
3588736
C1=CC(=CC=C1CCC(=O)O)C(F)(F)F
C10H9F3O2
MFCD00674032
4-trifluoromethyl hydrocinnamic acid, 3-4-trifluoromethyl phenyl propanoic acid, 3-4-trifluoromethyl phenyl propionic acid, benzenepropanoic acid, 4-trifluoromethyl, 3-4-trifluoromethylphenyl propionic acid, 4-trifluoromethyl phenylpropionic acid, p-trifluoromethyl benzylacetic acid, 3-4-trifluoromethyl-phenyl-propionic acid
3-[4-(trifluoromethyl)phenyl]propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions