Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-Toluenedithiol, 95%

$64.55 - $208.97

Chemical Identifiers

Chemical Name or Material 3,4-Toluenedithiol
CAS 496-74-2
Molecular Formula C7H8S2
Formula Weight 156.26g/mol
MDL Number MFCD00004844
Beilstein 1635270
Merck Index 14,9531
Synonym 3,4-Dimercaptotoluene; 1,2-Dimercapto-4-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1500303

 
alfa aesar™
L15003-03
1g Each for $64.55

AAL1500306

 
alfa aesar™
L15003-06
5g Each for $208.97
Description & Specifications

Specifications

Melting Point 28° to 33°C
Boiling Point 126° to 128°C (10mmHg)
Quantity 1g
Flash Point >110°C (230°F)
Odor Stench
Chemical Name or Material 3,4-Toluenedithiol
CAS 496-74-2
Assay 95%
Molecular Formula C7H8S2
Formula Weight 156.26g/mol
MDL Number MFCD00004844
Beilstein 1635270
Merck Index 14,9531
Synonym 3,4-Dimercaptotoluene; 1,2-Dimercapto-4-methylbenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
RTECS Number XT2275000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C