missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(4-tert-Butylphenyl)propionic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5401114

 View more versions of this product

Catalog No. AAH5401114



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-(4-tert-Butylphenyl)propionic acid
102°C
MFCD00796482
3-4-tert-butyl-phenyl-propionic acid, 3-4-tert-butyl phenyl propanoic acid, 3-4-tert-butylphenyl propanoic acid, 4-1,1-dimethylethyl benzenepropanoic acid, 4-t-butylhydrocinnamic acid, 4-t-butylhydrocinnamic aicd, 4-tert-butylhydrocinnamic acid, 3-4-t-butylphenyl propionic acid, 3-4-t-butylphenyl propanoic acid
CC(C)(C)C1=CC=C(C=C1)CCC(=O)O
206.285
206.28
1208-64-6
C13H18O2
25 g
BNJYANVQFVSYEK-UHFFFAOYSA-N
3-(4-tert-butylphenyl)propanoic acid
4962173
97%

Chemical Identifiers

1208-64-6
206.285
BNJYANVQFVSYEK-UHFFFAOYSA-N
4962173
CC(C)(C)C1=CC=C(C=C1)CCC(=O)O
C13H18O2
MFCD00796482
3-4-tert-butyl-phenyl-propionic acid, 3-4-tert-butyl phenyl propanoic acid, 3-4-tert-butylphenyl propanoic acid, 4-1,1-dimethylethyl benzenepropanoic acid, 4-t-butylhydrocinnamic acid, 4-t-butylhydrocinnamic aicd, 4-tert-butylhydrocinnamic acid, 3-4-t-butylphenyl propionic acid, 3-4-t-butylphenyl propanoic acid
3-(4-tert-butylphenyl)propanoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only