Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Piperidinyloxy)benzonitrile hydrochloride, 97%

$90.81 - $326.32

Chemical Identifiers

Chemical Name or Material 3-(4-Piperidinyloxy)benzonitrile hydrochloride
CAS 950649-07-7
Molecular Formula C12H14N2O·HCl
Formula Weight 238.7
MDL Number MFCD11977714
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5114303

  SDS
alfa aesar™
H5114303
1g Each for $326.32

AAH51143MD

  SDS
alfa aesar™
H51143MD
250mg Each for $90.81
Description & Specifications

Specifications

Formula Weight 238.7
Chemical Name or Material 3-(4-Piperidinyloxy)benzonitrile hydrochloride
Assay 97%
Molecular Formula C12H14N2O·HCl
MDL Number MFCD11977714
CAS 950649-07-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.