missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3,4'-Oxydiphthalic Anhydride 98.0+%, TCI America™
$36.49 - $404.79
Chemical Identifiers
CAS | 50662-95-8 |
---|---|
Molecular Formula | C16H6O7 |
Molecular Weight (g/mol) | 310.22 |
MDL Number | MFCD22460472 |
InChI Key | OPVHOFITDJSMOD-UHFFFAOYSA-N |
PubChem CID | 14574292 |
IUPAC Name | 4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]-1,3-dihydro-2-benzofuran-1,3-dione |
SMILES | O=C1OC(=O)C2=C1C=CC(OC1=CC=CC3=C1C(=O)OC3=O)=C2 |
Chemical Identifiers
50662-95-8 | |
310.22 | |
OPVHOFITDJSMOD-UHFFFAOYSA-N | |
4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]-1,3-dihydro-2-benzofuran-1,3-dione |
C16H6O7 | |
MFCD22460472 | |
14574292 | |
O=C1OC(=O)C2=C1C=CC(OC1=CC=CC3=C1C(=O)OC3=O)=C2 |
Specifications
50662-95-8 | |
White-Yellow | |
MFCD22460472 | |
OPVHOFITDJSMOD-UHFFFAOYSA-N | |
4-[(1,3-dioxo-1,3-dihydro-2-benzofuran-5-yl)oxy]-1,3-dihydro-2-benzofuran-1,3-dione | |
14574292 | |
≥98.0% (GC,T) | |
3,4′-Oxydiphthalic Anhydride |
180°C | |
C16H6O7 | |
1 g | |
O=C1OC(=O)C2=C1C=CC(OC1=CC=CC3=C1C(=O)OC3=O)=C2 | |
310.22 | |
310.22 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer