Please login to your online account to display your discounted pricing

Alfa Aesar™ 3,4-O-Isopropylidene-D-mannitol, 97%

$50.47 - $201.91

Chemical Identifiers

Chemical Name or Material 3,4-O-Isopropylidene-D-mannitol
CAS 3969-84-4
Molecular Formula C9H18O6
Formula Weight 222.24
MDL Number MFCD00075122
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2107003

  SDS
alfa aesar™
B2107003
Each for $50.47

AAB2107006

  SDS
alfa aesar™
B2107006
Each for $201.91
Description & Specifications

Specifications

Formula Weight 222.24
Chemical Name or Material 3,4-O-Isopropylidene-D-mannitol
Quantity 1g
Melting Point 85° to 88°C
Molecular Formula C9H18O6
Assay 97%
CAS 3969-84-4
MDL Number MFCD00075122