missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Morpholinylsulfonyl)benzeneboronic acid, 96%

Manufacturer:  Alfa Aesar™ H5324203

 View more versions of this product

Catalog No. AAH5324203



Specifications

Specifications

3-(4-Morpholinylsulfonyl)benzeneboronic acid
1g
C10H14BNO5S
3-morpholin-4-ylsulphonyl benzeneboronic acid, 3-morpholinosulfonyl phenylboronic acid, 3-morpholinosulfonyl phenyl boronic acid, 3-n-morpholinylsulfonylphenylboronic acid, 3-morpholine-4-sulfonyl phenylboronic acid, 3-n-morpholinylsulfonamidophenyl boronic acid, 4-3-boronophenyl sulphonyl morpholine, 3-morpholin-4-ylsulfonylphenyl boronic acid, 4-3-boronobenzenesulfonyl morpholine, 3-morpholine-4-sulfonyl phenyl boronic acid
B(C1=CC(=CC=C1)S(=O)(=O)N2CCOCC2)(O)O
271.094
271.09
144°C to 148°C
871329-60-1
MFCD07363748
HIKVVOUXXNJGAK-UHFFFAOYSA-N
(3-morpholin-4-ylsulfonylphenyl)boronic acid
44119633
96%

Chemical Identifiers

871329-60-1
271.094
HIKVVOUXXNJGAK-UHFFFAOYSA-N
44119633
B(C1=CC(=CC=C1)S(=O)(=O)N2CCOCC2)(O)O
C10H14BNO5S
MFCD07363748
3-morpholin-4-ylsulphonyl benzeneboronic acid, 3-morpholinosulfonyl phenylboronic acid, 3-morpholinosulfonyl phenyl boronic acid, 3-n-morpholinylsulfonylphenylboronic acid, 3-morpholine-4-sulfonyl phenylboronic acid, 3-n-morpholinylsulfonamidophenyl boronic acid, 4-3-boronophenyl sulphonyl morpholine, 3-morpholin-4-ylsulfonylphenyl boronic acid, 4-3-boronobenzenesulfonyl morpholine, 3-morpholine-4-sulfonyl phenyl boronic acid
(3-morpholin-4-ylsulfonylphenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents