Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Morpholinylmethyl)indole, 95%

Chemical Identifiers

Chemical Name or Material 3-(4-Morpholinylmethyl)indole
CAS 5379-88-4
Formula Weight 216.28
MDL Number MFCD00458413
Beilstein 193005
Synonym 4-(3-Indolylmethyl)morpholine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6336603

  SDS
alfa aesar™
H6336603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6336606

  SDS
alfa aesar™
H6336606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63366MD

  SDS
alfa aesar™
H63366MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.28
Chemical Name or Material 3-(4-Morpholinylmethyl)indole
Physical Form Solid
Assay 95%
MDL Number MFCD00458413
CAS 5379-88-4
Beilstein 193005
Synonym 4-(3-Indolylmethyl)morpholine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White to off-white
Quantity 1g
RTECS Number NM1140000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.