missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Morpholinylmethyl)indole, 95%

Manufacturer:  Alfa Aesar™ H6336606

 View more versions of this product

Catalog No. AAH6336606



Specifications

Specifications

3-(4-Morpholinylmethyl)indole
5g
C13H16N2O
193005
VHGISLOQNDQFIW-UHFFFAOYSA-N
4-(1H-indol-3-ylmethyl)morpholine
21477
Solid
White
5379-88-4
MFCD00458413
3-morpholinomethyl indole, indole, 3-morpholinomethyl, 3-morpholin-4-ylmethyl-1h-indole, 3-morpholin-4-ylmethyl-1h-indole, 4-27-00-00551 beilstein handbook reference, 4-indol-3-ylmethyl morpholine, 3-morpholinomethyl-1h-indole, morpholine, n-3-indolyl methyl, 4-1h-indol-3-ylmethyl morpholine, 3-4-morpholinylmethyl indole
C1COCCN1CC2=CNC3=CC=CC=C32
216.284
216.28
95%

Chemical Identifiers

5379-88-4
216.284
VHGISLOQNDQFIW-UHFFFAOYSA-N
21477
C1COCCN1CC2=CNC3=CC=CC=C32
C13H16N2O
MFCD00458413
3-morpholinomethyl indole, indole, 3-morpholinomethyl, 3-morpholin-4-ylmethyl-1h-indole, 3-morpholin-4-ylmethyl-1h-indole, 4-27-00-00551 beilstein handbook reference, 4-indol-3-ylmethyl morpholine, 3-morpholinomethyl-1h-indole, morpholine, n-3-indolyl methyl, 4-1h-indol-3-ylmethyl morpholine, 3-4-morpholinylmethyl indole
4-(1H-indol-3-ylmethyl)morpholine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NM1140000

SDS Documents