missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester

$47.86 - $743.47

Chemical Identifiers

CAS 1036991-25-9
Molecular Formula C17H24BNO4
Formula Weight 317.19
MDL Number MFCD09266172
Synonym 4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Chemical Name or Material 3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5111303 SDS Alfa Aesar™
H5111303
1g Each for $47.86
AAH5111306 SDS Alfa Aesar™
H5111306
5g Each for $183.18
AAH5111314 SDS Alfa Aesar™
H5111314
25g Each for $743.47
Specifications Description & Specifications

Chemical Identifiers

1036991-25-9
317.19
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
C17H24BNO4
MFCD09266172
3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester

Specifications

1036991-25-9
317.19
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
3-(4-Morpholinylcarbonyl)benzeneboronic acid pinacol ester
C17H24BNO4
MFCD09266172
1g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning