missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Morpholinyl)propyl isothiocyanate, 96%

Manufacturer:  Alfa Aesar™ L1281903

 View more versions of this product

Catalog No. AAL1281903



Specifications

Specifications

3-(4-Morpholinyl)propyl isothiocyanate
1g
C8H14N2OS
UN2922
3-morpholinopropyl isothiocyanate, 4-3-isothiocyanatopropyl morpholine, 3-4-morpholino propyl isothiocyanate, 3-morpholinopropylisothiocyanate, 3-morpholin-4-ylpropanisothiocyanate, 3-4-morpholinyl propyl isothiocyanate, 3-morpholin-4-yl propylisothiocyanate, 4-3-isothiocyanato-propyl-morpholine, 4-3-isothiocyanatopropyl morpholine #, morpholine,4-3-isothiocyanatopropyl
C1COCCN1CCCN=C=S
186.273
186.28
182°C to 183°C (13mmHg)
32813-50-6
MFCD00041222
879217
BCEFDMYFAAAFPE-UHFFFAOYSA-N
4-(3-isothiocyanatopropyl)morpholine
141745
96%

Chemical Identifiers

32813-50-6
186.273
BCEFDMYFAAAFPE-UHFFFAOYSA-N
141745
C1COCCN1CCCN=C=S
C8H14N2OS
MFCD00041222
3-morpholinopropyl isothiocyanate, 4-3-isothiocyanatopropyl morpholine, 3-4-morpholino propyl isothiocyanate, 3-morpholinopropylisothiocyanate, 3-morpholin-4-ylpropanisothiocyanate, 3-4-morpholinyl propyl isothiocyanate, 3-morpholin-4-yl propylisothiocyanate, 4-3-isothiocyanato-propyl-morpholine, 4-3-isothiocyanatopropyl morpholine #, morpholine,4-3-isothiocyanatopropyl
4-(3-isothiocyanatopropyl)morpholine
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Recommended Storage : 2° to 8°C

Documents