Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Morpholinyl)propiophenone hydrochloride, 98+%

$99.89 - $322.45

Chemical Identifiers

Chemical Name or Material 3-(4-Morpholinyl)propiophenone hydrochloride
CAS 1020-16-2
Molecular Formula C13H17NO2·HCl
Formula Weight 255.76
MDL Number MFCD00035332
Beilstein 3723608
Synonym 3-(4-Morpholino)propiophenone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2368614

  SDS
alfa aesar™
B2368614
25g Each for $99.89

AAB2368622

  SDS
alfa aesar™
B2368622
100g Each for $322.45
Description & Specifications

Specifications

Formula Weight 255.76
Chemical Name or Material 3-(4-Morpholinyl)propiophenone hydrochloride
Assay ≥98%
Molecular Formula C13H17NO2·HCl
MDL Number MFCD00035332
CAS 1020-16-2
Beilstein 3723608
Synonym 3-(4-Morpholino)propiophenone hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 180° to 182°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.