Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-(4-Methylsulfonylbenzoyl)propionic acid, 98%

$82.26 - $301.35

Chemical Identifiers

Chemical Name or Material 3-(4-Methylsulfonylbenzoyl)propionic acid
CAS 7028-79-7
Molecular Formula C11H12O5S
Formula Weight 256.28
MDL Number MFCD00082738
Beilstein 2134110
Synonym 3-(4-Methanesulfonylbenzoyl)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1275003

  SDS
alfa aesar™
L1275003
1g Each for $82.26

AAL1275006

  SDS
alfa aesar™
L1275006
5g Each for $301.35
Description & Specifications

Specifications

Formula Weight 256.28
Chemical Name or Material 3-(4-Methylsulfonylbenzoyl)propionic acid
Molecular Formula C11H12O5S
MDL Number MFCD00082738
Synonym 3-(4-Methanesulfonylbenzoyl)propionic acid
Health Hazard 1 Warning
Melting Point 168° to 172°C
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
CAS 7028-79-7
Beilstein 2134110
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.